Anti-RALGDS Rabbit Polyclonal Antibody

Lieferant: Abgent
AP18831C
ABGEAP18831CEA 354 EUR
ABGEAP18831C
Anti-RALGDS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RALGDS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR