Anti-APBA3 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP18975C
ABGEAP18975CEA 354 EUR
ABGEAP18975C
Anti-APBA3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: APBA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR