Anti-MCCC2 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP18978B
ABGEAP18978BEA 354 EUR
ABGEAP18978B
Anti-MCCC2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MCCC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human