Anti-TMPPE Rabbit Polyclonal Antibody

Lieferant: Abgent
AP19243C
ABGEAP19243CEA 354 EUR
ABGEAP19243C
Anti-TMPPE Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TMPPE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR