Anti-TMPRSS6 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP19372A
ABGEAP19372AEA 354 EUR
ABGEAP19372A
Anti-TMPRSS6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TMPRSS6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR