Anti-EIF3E Rabbit Polyclonal Antibody

Lieferant: Abgent
AP19409C
ABGEAP19409CEA 354 EUR
ABGEAP19409C
Anti-EIF3E Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EIF3E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human, Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR