Anti-EOMES Rabbit Polyclonal Antibody
Lieferant: Abgent
AP19703A
ABGEAP19703AEA 354 EUR
ABGEAP19703A
Anti-EOMES Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EOMES
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human