Anti-ACKR1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP20261A
ABGEAP20261AEA 354 EUR
ABGEAP20261A
Anti-ACKR1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ACKR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR