Anti-EDC3 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP20283C
ABGEAP20283CEA 354 EUR
ABGEAP20283C
Anti-EDC3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EDC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human