Anti-LDB3 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP20668A
ABGEAP20668AEA 354 EUR
ABGEAP20668A
Anti-LDB3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LDB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Mouse