Anti-H2AFX Rabbit Polyclonal Antibody
Lieferant: Abgent
AP20702B
ABGEAP20702BEA 354 EUR
ABGEAP20702B
Anti-H2AFX Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: H2AFX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human