Anti-H2AFX Rabbit Polyclonal Antibody

Lieferant: Abgent
AP20703B
ABGEAP20703BEA 354 EUR
ABGEAP20703B
Anti-H2AFX Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: H2AFX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR