Anti-AR Rabbit Polyclonal Antibody

Lieferant: Abgent
AP2509A
ABGEAP2509AEA 354 EUR
ABGEAP2509A
Anti-AR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AR (androgen receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR