Anti-MARCKS Rabbit Polyclonal Antibody

Lieferant: Abgent
AP2521B
ABGEAP2521BEA 354 EUR
ABGEAP2521B
Anti-MARCKS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MARCKS (myristoylated alanine-rich protein kinase C substrate)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR