Anti-AKR1A1 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP2734B
ABGEAP2734BEA 354 EUR
ABGEAP2734B
Anti-AKR1A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AKR1A1 (aldo-keto reductase family 1, member A1 (aldehyde reductase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human