Anti-FABP4 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP2764D
ABGEAP2764DEA 354 EUR
ABGEAP2764D
Anti-FABP4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FABP4 (fatty acid binding protein 4, adipocyte)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR