Anti-U2AF1 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP2802D
ABGEAP2802DEA 354 EUR
ABGEAP2802D
Anti-U2AF1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: U2AF1 (U2 small nuclear RNA auxiliary factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human