Anti-STRAP Rabbit Polyclonal Antibody
Lieferant: Abgent
AP2933B
ABGEAP2933BEA 354 EUR
ABGEAP2933B
Anti-STRAP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STRAP (serine/threonine kinase receptor associated protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human