Anti-GOT1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP2947A
ABGEAP2947AEA 354 EUR
ABGEAP2947A
Anti-GOT1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GOT1 (glutamic-oxaloacetic transaminase 1, soluble)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR