Anti-BRAF Rabbit Polyclonal Antibody

Lieferant: Abgent
AP3513A
ABGEAP3513AEA 354 EUR
ABGEAP3513A
Anti-BRAF Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BRAF (v-raf murine sarcoma viral oncogene homolog B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR