Anti-PAFAH1B2 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP3773A
ABGEAP3773AEA 354 EUR
ABGEAP3773A
Anti-PAFAH1B2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PAFAH1B2 (platelet-activating factor acetylhydrolase 1b, catalytic subunit 2 (30kDa))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR