Anti-PTEN Rabbit Polyclonal Antibody

Lieferant: Abgent
AP3788A
ABGEAP3788AEA 354 EUR
ABGEAP3788A
Anti-PTEN Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTEN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR