Anti-RELA Rabbit Polyclonal Antibody

Lieferant: Abgent
AP3861A
ABGEAP3861AEA 354 EUR
ABGEAP3861A
Anti-RELA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RELA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR