Anti-CDKN1B Rabbit Polyclonal Antibody

Lieferant: Abgent
AP3874A
ABGEAP3874AEA 354 EUR
ABGEAP3874A
Anti-CDKN1B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDKN1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR