Anti-F1LVT0 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP3880A
ABGEAP3880AEA 354 EUR
ABGEAP3880A
Anti-F1LVT0 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: F1LVT0
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR