Anti-TSC1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP3894A
ABGEAP3894AEA 354 EUR
ABGEAP3894A
Anti-TSC1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TSC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR