Anti-ISLR Rabbit Polyclonal Antibody

Lieferant: Abgent
AP4877B
ABGEAP4877BEA 354 EUR
ABGEAP4877B
Anti-ISLR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ISLR (immunoglobulin superfamily containing leucine-rich repeat)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR