Anti-KCTD1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP4955B
ABGEAP4955BEA 354 EUR
ABGEAP4955B
Anti-KCTD1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KCTD1 (potassium channel tetramerization domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR