Anti-CASP3 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP4985D
ABGEAP4985DEA 354 EUR
ABGEAP4985D
Anti-CASP3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CASP3 (caspase 3, apoptosis-related cysteine peptidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR