Anti-KRT82 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP5049C
ABGEAP5049CEA 354 EUR
ABGEAP5049C
Anti-KRT82 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KRT82 (keratin 82)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR