Anti-RPS26 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP5261A
ABGEAP5261AEA 354 EUR
ABGEAP5261A
Anti-RPS26 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RPS26 (ribosomal protein S26)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human