Anti-UBAC2 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP5292A
ABGEAP5292AEA 354 EUR
ABGEAP5292A
Anti-UBAC2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UBAC2 (UBA domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR