Anti-ZBTB5 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP5302A
ABGEAP5302AEA 354 EUR
ABGEAP5302A
Anti-ZBTB5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZBTB5 (zinc finger and BTB domain containing 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR