Anti-GNE Rabbit Polyclonal Antibody

Lieferant: Abgent
AP5318B
ABGEAP5318BEA 354 EUR
ABGEAP5318B
Anti-GNE Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GNE (glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR