Anti-AAGAB Rabbit Polyclonal Antibody

Lieferant: Abgent
AP5483B
ABGEAP5483BEA 354 EUR
ABGEAP5483B
Anti-AAGAB Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AAGAB (alpha- and gamma-adaptin binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR