Anti-BACE2C Rabbit Polyclonal Antibody

Lieferant: Abgent
AP6106A
ABGEAP6106AEA 354 EUR
ABGEAP6106A
Anti-BACE2C Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BACE2C
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR