Anti-NRG2 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP6223A
ABGEAP6223AEA 354 EUR
ABGEAP6223A
Anti-NRG2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NRG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (350–400 and 800–850 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR