Anti-Presenilin 1 (PSEN1) Rabbit Polyclonal Antibody

Lieferant: Abgent
AP6231A
ABGEAP6231AEA 354 EUR
ABGEAP6231A
Anti-Presenilin 1 (PSEN1) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Presenilin 1 (PSEN1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR