Anti-DDIT4 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP6268A
ABGEAP6268AEA 354 EUR
ABGEAP6268A
Anti-DDIT4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DDIT4 (DNA-damage-inducible transcript 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR