Anti-CLDN1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP6308B
ABGEAP6308BEA 354 EUR
ABGEAP6308B
Anti-CLDN1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CLDN1 (claudin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: Loop 2
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR