Anti-Park6 (PINK1) Rabbit Polyclonal Antibody

Lieferant: Abgent
AP6406D
ABGEAP6406DEA 354 EUR
ABGEAP6406D
Anti-Park6 (PINK1) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Park6 (PINK1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR