Anti-CPB2 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP6541C
ABGEAP6541CEA 354 EUR
ABGEAP6541C
Anti-CPB2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CPB2 (carboxypeptidase B2 (plasma))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR