Anti-TIMP1 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP6709A
ABGEAP6709AEA 354 EUR
ABGEAP6709A
Anti-TIMP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TIMP1 (TIMP metallopeptidase inhibitor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human