Anti-FUCA1 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP6791C
ABGEAP6791CEA 354 EUR
ABGEAP6791C
Anti-FUCA1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FUCA1 (fucosidase, alpha-L- 1, tissue)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR