Anti-SAR1A Rabbit Polyclonal Antibody

Lieferant: Abgent
AP6915C
ABGEAP6915CEA 354 EUR
ABGEAP6915C
Anti-SAR1A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SAR1A (secretion associated, Ras related GTPase 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR