Anti-AURKC Rabbit Polyclonal Antibody

Lieferant: Abgent
AP7000G
ABGEAP7000GEA 354 EUR
ABGEAP7000G
Anti-AURKC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AURKC (aurora kinase C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR