Anti-PRKAB2 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP7046A
ABGEAP7046AEA 354 EUR
ABGEAP7046A
Anti-PRKAB2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRKAB2 (protein kinase, AMP-activated, beta 2 non-catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse