Anti-Creatine kinase MT (CKMT1) Rabbit Polyclonal Antibody

Lieferant: Abgent
AP7071A
ABGEAP7071AEA 354 EUR
ABGEAP7071A
Anti-Creatine kinase MT (CKMT1) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Creatine kinase MT (CKMT1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR