Anti-PNCK Rabbit Polyclonal Antibody

Lieferant: Abgent
AP7097A
ABGEAP7097AEA 354 EUR
ABGEAP7097A
Anti-PNCK Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PNCK (pregnancy up-regulated nonubiquitous CaM kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR