Anti-GK2 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP7131B
ABGEAP7131BEA 354 EUR
ABGEAP7131B
Anti-GK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GK2 (glycerol kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR